Learn More
Agilent Technologies Storage/reaction microplate, 24-well, irradiated polypropylene, 25/pk
Supplier:  Agilent Technologies 204023100
Encompass
Storage/reaction microplate, 24-well, irradiated polypropylene, 10 mL/square well, pyramid bottoms, 44 mm height, individually bagged, 25/pk.
Catalog No. NC1130477