Learn More
Optimize Technologies Small Seal Forming/Insertion Tool
SDP
Supplier:  Optimize Technologies 101002699
Encompass_Preferred
Installing a piston seal can be a tricky undertaking. A relatively small seal must fit into the seal cavity in such a way to ensure the seal is inserted evenly without distortion or damage. The Optimize Seal Forming/Insertion Tool Kit is specifically designed for easy and successful installation. One end has a beveled cavity for seal forming while the other end is an insertion support for perfect seal installation. Each kit contains instructions for maximizing seal life and a Solvent Compatibility Guide to aid in seal material selection.
Catalog No. 50-113-285