Learn More
Ted Pella Inc SEAL N FREEZE LARGE 50/PK
SDP
Supplier:  Ted Pella Inc 209562
Encompass
SEAL'N FREEZE® CRYOTRAY®Cryo Embedding Molds.-- Large: 25 L x 30 W x 20mm D (1 L x 1.2 W x 0.8" D), pkg/50 trays (200 molds). The SEAL’N FREEZE® CRYOTRAY® is a compact system of plastic cryo embedding molds with attached sealable lids. This tray system allows for batch processing of tissue samples in perfect uniform blocks that are ideal for cryo sectioning and storage.Each disposable plastic SEAL’N FREEZE® CRYOTRAY® includes four individual molds which can be easily separated from the center connector by folding and tearing along the perforated borders.
Catalog No. NC2052005