Learn More
Research Products International Corp SealPlate ColorTab Film, Non-S, Orange, 100/pk
SDP
Supplier:  Research Products International Corp 202527O
Encompass
These films feature a color dot and white writing area on each end tab for convenient plate identification. End tab can be left intact even if center of film is removed from plate. Second end tab can be marked and separated for safe keeping. Clear adhesive film prevents sample evaporation. SealPlate ColorTab is a Trademark of Excel Scientific, Inc.
Catalog No. 50-223-0143