Learn More
Medchemexpress LLC Sebutylazine-dd5 (ethyl-dd5) | 1219805-56-7 | C9H11D5ClN5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY151913S5MG
Encompass_Preferred
Sebutylazine-dd5 (ethyl-dd5) is a deuterium labeled chemical intended for research use only. This compound serves as a tracer and an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS. Its deuterium labeling can impact pharmacokinetic and metabolic profiles.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • For research use only
Catalog No. 50-003-72797