Learn More
HF Scientific Inc ProCal Primary Calibration Kit, Full Range, .02, 10, and 1000 NTU
Supplier:  HF Scientific Inc 39957
Encompass
ProCal Primary Calibration Kit .02 & 10 & 1000 NTU (for MicroTOL 2 and MicroTOL4 turbidimeters). HF Scientific ProCal turbidity standard and calibration kit. ProCal turbidity standards are pre-diluted, factory certified, and ready for use with not only the HF scientific turbidimeter product line but with other manufacturers instrumentation. The ProCal standard formulation utilizes a preservative that enhances stability, yet is safe and biodegrades rapidly in aquatic environments.
Catalog No. 15-393-101