Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp SEC Column, 125 Å Pore, MaxPeak Premier, 1.7 μm, 4.6 x 300 mm

Supplier:  Waters Corp 186011351

Encompass_Preferred

This SEC column is developed for scientists working with biotherapeutic peptides and small proteins (1K - 80K Dalton). It provides higher resolution of size variants, more reproducible HMWPs quantitation, reduced run times, and solvent cost savings. It utilizes MaxPeak High Performance Surface (HPS) Technologies to minimize non-desired secondary ionic interactions.

  • Particle substrate: Hybrid
  • pH range: 2.5 to 8
  • Temperature limits: 60 °C
  • Maximum pressure: 11,000 psi (758 bar)
  • Bonding technology: Diol (OH)
  • Silanol activity: Low
  • Molecular weight range: 1,000 to 80,000
  • Particle shape: Spherical
  • Particle size: 1.7 μm
  • Endfitting type: Parker-style
  • Pore size: 125 Å
  • QC tested: Peptide and protein
  • System compatibility: UPLC, UHPLC

Catalog No. NC3887720


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.