Learn More
Medchemexpress LLC HY-123972 1mg Medchemexpress, SEC inhibitor KL-2 CAS:900308-51-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1239721MG
Encompass_Preferred
Medchemexpress, HY-123972 1mg SEC inhibitor KL-2 CAS:900308-51-2 SEC inhibitor KL-2 (KL-2), a peptidomimetic lead compound, is a potent, selective super elongation complex (SEC) inhibitor and disrupts the interaction between the SEC scaffolding protein AFF4 and P-TEFb, resulting in impaired release of Pol II from promoter-proximal pause sites and a reduced average rate of processive transcription elongation. SEC inhibitor KL-2 exhibits an dose-dependent inhibitory effect on AFF4-CCNT1 interaction with a Ki of 1.50 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9655