Learn More
ABclonal Technology Sec23B Rabbit pAb
SDP
Supplier:  ABclonal Technology A151311000UL
Encompass_Preferred
The protein encoded by this gene is a member of the SEC23 subfamily of the SEC23/SEC24 family, which is involved in vesicle trafficking. The encoded protein has similarity to yeast Sec23p component of COPII. COPII is the coat protein complex responsible for vesicle budding from the ER. The function of this gene product has been implicated in cargo selection and concentration. Multiple alternatively spliced transcript variants have been identified in this gene.
Catalog No. 50-001-09252