Learn More
Medchemexpress LLC Seco-DUBA | 1227961-59-2 | 98.1% | 526.97 | 1 MG
SDP
Supplier:  Medchemexpress LLC 1MGHY132180A
Encompass_Preferred
Seco-DUBA is a duocarmycin (DUBA) prodrug that contains two hydroxyl groups, each capable of coupling to an antibody via a linker. It is primarily used in the synthesis of antibody-drug conjugates (ADCs). Research indicates that Seco-DUBA dose-dependently reduces cell viability, demonstrating potency and efficacy comparable to DUBA in SK-BR-3 cells. It also induces high sensitivity in SK-BR-3, SK-OV-3, and SW620 cell lines. In vivo studies suggest that Seco-DUBA is likely converted to DUBA almost instantaneously.
  • Contains two hydroxyl groups for antibody coupling via a linker
  • Used in the synthesis of antibody-drug conjugates (ADCs)
  • Dose-dependently reduces cell viability
  • Potent and efficacious as DUBA in SK-BR-3 cells
  • Causes high sensitivity in SK-BR-3, SK-OV-3, and SW620 cells
  • Likely converts to DUBA almost instantaneously in vivo
Catalog No. 50-003-65470