Learn More
Medchemexpress LLC Seco-DUBA | 1227961-59-2 | 98.1% | 526.97 | C29H23ClN4O4 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY132180A25MG
Encompass_Preferred
Seco-DUBA is a duocarmycin (DUBA) prodrug containing two hydroxyl groups that enable coupling to an antibody via a linker. It is intended as an ADC payload for synthesis and study of antibody-drug conjugates.
  • A duocarmycin-based prodrug designed for ADC applications.
  • Contains two hydroxyl groups suitable for antibody coupling via a linker.
  • Used in the synthesis of antibody-drug conjugates for targeted delivery studies.
  • Exhibits dose-dependent cytotoxicity comparable to DUBA.
  • Demonstrates high in vitro potency against multiple cancer cell lines (reported IC50 values).
Catalog No. 50-002-03700
Add to Cart
Request a Quote