Learn More
Medchemexpress LLC Seco rapamycin ethyl ester | 00-00-0 | 85.1% | C53H83NO13 | 10MG

Supplier: Medchemexpress LLC HY13377010MG
Seco Rapamycin ethyl ester is an open-ring metabolite of rapamycin derivatives supplied for research use. It is reported not to affect mTOR function and is provided as a light yellow oil (molecular formula C53H83NO13). Typical packaging is small milligram quantities for laboratory studies; confirm storage and purity with accompanying documentation.
- Open-ring metabolite of rapamycin derivative.
- Reported not to affect mTOR function.
- Molecular formula C53H83NO13; molecular weight 942.23.
- Light yellow oil appearance; store at -20°C and protect from light.
- Available in small milligram pack sizes for research use.
- Purity varies by source; confirm certificate of analysis before use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.