Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (+)-Secoisolariciresinol | 145265-02-7 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYN817210MG

Encompass_Preferred

(+)-Secoisolariciresinol is a lignan compound and is the (+)-enantiomer of Secoisolariciresinol. This product is for research use only.

  • Purity of 99.66%
  • Molecular weight of 362.42
  • Chemical formula C20H26O6
  • Solid appearance
  • White to off-white color
  • Initial source from plants (Leguminosae, Caesalpinia magnifoliolata Metc. and Compositae)
  • Ships at room temperature in continental US; may vary elsewhere
  • Storage at 4°C, sealed, away from moisture and light. In solvent: -80°C for 6 months; -20°C for 1 month (sealed, away from moisture and light)
  • In vitro solubility in DMSO: 50 mg/mL (137.96 mM); requires ultrasonic; hygroscopic DMSO has a significant impact on solubility, so use newly opened DMSO
  • In vivo solubility: protocols available for clear solution at >= 2.5 mg/mL

Catalog No. 50-004-19739


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.