Learn More
Biotium Secondary Antibody Biotin 3D6.6 CF405S 1/EA
Supplier:  Biotium 20203
Encompass_Preferred
This product is prepared by labeling monoclonal mouse anti-biotin with CF and reg 405S dye CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes Blue fluorescent CF and reg 405S dye has excitation/emission at 404/431 nm Note Conjugates of blue fluorescent dyes like CF and reg 350 CF and reg 405S and CF and reg 405M are not recommended for detecting low abundance targets because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors
Catalog No. 50-005-00202