Learn More
Biotium Secondary Antibody Dinitrophenyl DNP Rabbit CF750 1/EA
Supplier:  Biotium 2087550UL
Encompass_Preferred
High-quality rabbit anti-dinitrophenyl (DNP) labeled with near-IR CF750 dye (Ex/Em 755/777 nm) CF dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes 2 mg/mL 50 uL
Catalog No. 50-005-00211