Learn More
Biotium Secondary Antibody Goat IgG Donkey CF498 1/EA
Supplier:  Biotium 20992500UL
Encompass_Preferred
This product is prepared by labeling highly cross-adsorbed donkey anti-goat IgG (H plus L) with CF and reg 498 dye and has excitation/emission at 497/525 nm Single label antibodies have an average of one dye molecule per antibody molecule which is reported to be optimal for STORM imaging
Catalog No. 50-004-99719