Learn More
Biotium Secondary Antibody Goat IgG Rabbit CF583R 1/EA
Supplier:  Biotium 2091050UL
Encompass_Preferred
This product is prepared by labeling high quality rabbit anti-goat IgG (H plus L) with CF and reg 583R dye CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes
Catalog No. 50-005-00172