Learn More
Biotium Secondary Antibody Guinea Pig IgG Donkey CF583R 1/EA
Supplier:  Biotium 20893500UL
Encompass_Preferred
This product is prepared by labeling highly cross-adsorbed donkey anti-guinea pig IgG (H plus L) with CF and reg 583R dye To minimize cross-reactivity the antibody has been adsorbed against bovine chicken goat Syrian hamster horse human mouse rabbit sheep and rat serum
Catalog No. 50-005-00660