Learn More
Biotium Secondary Antibody Mouse IgG Donkey CF583R 1/EA
Supplier:  Biotium 2089450UL
Encompass_Preferred
This product is prepared by labeling highly cross-adsorbed donkey anti-mouse IgG (H plus L) with CF and reg 583R dye To minimize cross-reactivity the antibody has been adsorbed against bovine chicken goat guinea pig Syrian hamster horse human rabbit and sheep serum
Catalog No. 50-004-99474