Learn More
Biotium Secondary Antibody Mouse IgG Goat CF583R 1/EA
Supplier:  Biotium 20903500UL
Encompass_Preferred
This product is prepared by labeling highly cross-adsorbed goat anti-mouse IgG (H plus L) with CF and reg 583R dye To minimize crossreactivity the antibody has been adsorbed against bovine chicken goat guinea pig hamster horse human rabbit rat and sheep serum
Catalog No. 50-005-00677