Learn More
Biotium Secondary Antibody Rabbit IgG Goat CF583R 1/EA
Supplier:  Biotium 20913250UL
Encompass_Preferred
This product is prepared by labeling high quality F(ab)2 fragment of goat anti-rabbit IgG (H plus L) with CF and reg 583R dye CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes
Catalog No. 50-005-00136