Learn More
Biotium Secondary Antibody Rabbit IgG Rat IgG Llama CF583R 1/EA
Supplier:  Biotium 2090950UL
Encompass_Preferred
This product is prepared by labeling an affinity-purified llama anti-rabbit IgG with CF and reg 583R dye CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes
Catalog No. 50-005-01171