Learn More
Medchemexpress LLC HY-N3009 20mg Medchemexpress, Secoxyloganin CAS:58822-47-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN300920MG
Encompass_Preferred
Medchemexpress, HY-N3009 20mg Secoxyloganin CAS:58822-47-2 Secoxyloganin, isolated from Lonicera japonica Thunb, inhibits the blood flow (BF) decrease. Secoxyloganin has allergy-preventive activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2912