Learn More
Medchemexpress LLC β-Secretase Inhibitor IV | 797035-11-1 | 99.2% | 578.72 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY10133
Encompass_Preferred
β-Secretase Inhibitor IV is a potent, cell-active BACE-1 inhibitor. It inhibits the production of Aβ peptides and suppresses the secretion of sAPP_NF and EV40 from HEK293T cells. This product is intended for research use only.
  • Potent BACE-1 inhibitor
  • Cell-active compound
  • Inhibits production of Aβ peptides
  • Suppresses secretion of sAPP_NF and EV40
  • Targets beta-secretase
  • Involved in neuronal signaling pathway
Catalog No. 50-003-47314