Learn More
Cayman Chemical Secreted AlkalIn Phosphatse
SDP
Supplier:  Cayman Chemical 600260300 TEST
Encompass_Preferred
SEAP is commonly used as a reporter of gene expression. Compared to other conventional intracellular reporters such as CAT and firefly luciferase, SEAP has the advantage of being secreted from transfected cells into the culture medium. Cayman’s Secreted Alkaline Phosphatase Reporter Gene Assay Kit (Luminescence) provides a simple chemiluminescence method for the sensitive quantitation of SEAP in conditioned cell culture medium from transfected cells.
Catalog No. 50-289-2332