Learn More
CPC Scientific H-His-Ser-Asp-Gly-Thr-Phe-Thr-Ser-Glu-Leu-Ser-Arg-Leu-Arg-Asp-Ser-Ala-Arg-Leu-Gln-Arg-Leu-Leu-Gln-Gly-Leu-Val-NH2 (acetate salt) 1MG
SDP
Supplier:  CPC Scientific SECT003B
Encompass
ONE-LETTER SEQUENCE: HSDGTFTSELSRLRDSARLQRLLQGLV-NH2MOLECULAR FORMULA: C130H220N44O41MOLECULAR WEIGHT:3055.45STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [17034-34-3], RESEARCH AREA: GastrointestinalREFERENCES: M.Hoshino et al., FEBS Letters, 178, 233 (1984); J. Liu et al., Nature Med<
Catalog No. 50-211-0391