Learn More
Electron Microscopy Sciences Section Lifter Model B
SDP
Supplier:  Electron Microscopy Sciences 6307620
Encompass_Preferred
Made from polished stainless steel. Perforated blade measures: 2.5"x2" (6.4x5.1cm), overall length is 7" (17.8cm)
Catalog No. 50-285-38