Learn More
Selleck Chemical LLC Securinine S9249-1MG
SDP
Supplier:  Selleck Chemical LLC S92491MG
Encompass_Preferred
Securinine a major natural alkaloid product from the root of the plant Securinega suffruticosa acts as a -amino butyric acid (GABA) receptor antagonist
Catalog No. 50-000-23908