Learn More
Medchemexpress LLC (-)-Securinine | 5610-40-2 | MFCD00078775 | 99.9% | 217.27 g/mol | C13H15NO2 | 10 MG

Supplier: Medchemexpress LLC HYN207910MG
(-)-Securinine is a plant-derived alkaloid supplied for research use that functions as a GABAA receptor antagonist. It is used in pharmacology and neuroscience research to probe GABAergic signaling and receptor pharmacology.
- High purity (99.9%) suitable for analytical and research applications.
- Molecular weight 217.27 g/mol; formula C13H15NO2.
- Available in small solid quantities (for example, 10 MG) and as a 10 mM solution in DMSO.
- Light yellow crystalline solid; store at recommended temperatures for long-term stability.
- Soluble in DMSO (50 mg/mL with warming); DMSO hygroscopicity can affect solubility-use freshly opened solvent.
- Intended for research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.