Learn More
Medchemexpress LLC Sedoheptulose anhydride | 469-90-9 | MFCD00150523 | 99.9% | 192.17 g/mol | C7H12O6 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW1455005MG
Encompass_Preferred
Sedoheptulose anhydride is a seven-carbon carbohydrate derivative supplied as a solid powder for biochemical research. It is used as a reagent or metabolic intermediate in studies of carbohydrate metabolism and heptose pathways. This material is intended for research use only.
  • High purity (reported 99.93%) suitable for analytical and preparative work.
  • Specific molecular formula and weight: C7H12O6, 192.17 g/mol.
  • Stable as a powder when stored under recommended conditions.
  • Available in small package sizes appropriate for lab-scale experiments.
  • Includes storage and stability guidance for powder and solution forms.
Catalog No. 50-002-83769