Learn More
Bioworld Seeband Protein Stain, 500 mL
Supplier:  Bioworld 214700111
Encompass_Preferred
Blue staining gel similar to Coomassie staining for protein gel electrophoresis. Allows electro-elution of a desired protein with high recovery yield. No denaturative material affecting the protein during staining Guarantees protein staining in polyacrylamide gels with only slight fixation of the protein to the gel. Low background . No need for multi-step washing Washing uses water only Storage at 4ºC. Shelf life 12 months 500ml
Catalog No. 50-198-718