Learn More
Apexbio Technology LLC Seglitide acetate(Synonyms: Vapreotide acetate, Sanvar, RC-160 acetate, Octastatin acetate, BIM-23014 acetate), 1mg, CAS: 99248-33-6.
SDP
Supplier:  Apexbio Technology LLC B68711
Encompass_Preferred
Seglitide acetate (CAS 99248-33-6) is a selective agonist for somatostatin receptor subtype 2 (sst2) characterized by oral bioavailability Through activation of sst2 receptors it modulates endocrine functions notably influencing growth hormone release and glucagon secretion Experimental studies indicate that Seglitide acetate affects blood pressure regulation and glucose metabolism making it a useful tool in diabetes research to elucidate hormonal regulatory pathways and potential therapeutic targets in endocrine-related disorders
Catalog No. 50-000-61244