Learn More
Medchemexpress LLC sEH inhibitor-7 | 340221-20-7 | 99.5% | 247.33 | 25 MG

Supplier: Medchemexpress LLC 25MGHY148136
sEH inhibitor-7 (compound c-2) is an inhibitor of soluble epoxide hydrolase (sEH) with IC50s of 0.15 μM (mouse sEH) and 6.2 μM (human sEH). Soluble epoxide hydrolase (sEH) plays an important role in the metabolism of endogenous chemical mediators involved in the regulation of blood pressure and inflammation.
- Inhibitor of soluble epoxide hydrolase (sEH)
- IC50s of 0.15 μM (mouse sEH) and 6.2 μM (human sEH)
- Plays an important role in the metabolism of endogenous chemical mediators
- Involved in the regulation of blood pressure and inflammation
- Purity: 99.5%
- Appearance: solid
- Color: off-white to light yellow
- Molecular weight: 247.33
- CAS No.: 340221-20-7
- Solubility: 100 mg/mL in DMSO
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.