Learn More
Medchemexpress LLC Seladelpar (lysine dihydrate) | 928821-40-3 | 98.0% | 626.68 g·mol^-1 | C27H41F3N2O9S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY19522C10MG
Encompass_Preferred
Seladelpar lysine dihydrate is the lysine dihydrate salt form of seladelpar, a selective PPAR-δ agonist supplied as a white to off-white solid for preclinical and in vitro research. The material is characterized for identity and purity and is intended for laboratory studies of metabolic and cholestatic liver disease pathways.
  • Selective PPAR-δ agonist activity for mechanistic studies.
  • Supplied as a solid with consistent physical appearance.
  • High purity, typically 98.0%.
  • Identified by CAS number 928821-40-3.
  • Available in small research quantities for laboratory use.
Catalog No. 50-002-94857