Learn More
TARGETMOL CHEMICALS INC SELAMECTIN 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T405925MG
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Selamectin (UK-124114) is a macrocyclic lactone derivative of ivermectin an anthelmintic that potentiates glutamate- and GABA-gated chloride channel opening in nematodes. In an H. contortus larval development assay selamectin (0.1 ug/ml) shows the effect of growth inhibition. purity: 99%
Catalog No. 50-257-9981