Learn More
Echelon Biosciences Research Labs Select-HA Mega Ladder
SDP
Supplier:  Echelon Biosciences Research Labs HYAMGLAD20UG
Encompass
The Select-HA Mega Ladder is a mixture of biotin-Select HA molecules of defined sizes for use as size standards in gel electrophoresis or other separation methods. This ladder covers a range from 2 megadalton to 8 megadalton. Select-HA™ is a hyaluronic acid (HA) preparation of uniform and narrow size distribution prepared by in vitro synthesis using recombinant Pasteurella multocida hyaluronan synthase. This item is non returnable
Catalog No. NC1775814