Learn More
Norell Inc 5mm, 900MHz 7" SELECT SERIES NMR TUBE, 5 Tubes per pack
SDP
Supplier:  Norell Inc S59007
Encompass_Preferred
Select Series NMR Tubes are ideal for cold refrigeration storage, work with cryo-probes and, variable temperature studies. Key properties that make this glass type desirable for NMR are its high degree of thermal shock resistance and low expansion coefficient. This allows for a greater margin of safety from breakage when used in variable temperature applications and freeze/thaw cycling, or under any other application where large temperature variations are required in the experiment.
Catalog No. NC2851731