Learn More
Molecular Dimensions SELENO MEDIA KIT
SDP
Supplier:  Molecular Dimensions MD12500
Encompass
Protein Crystallography. Based on a synthetic M9 minimal media supplemented with glucose, vitamins and amino acids with the exception of L-methionine. SelenoMethionine Medium Complete: Synthetic M9 minimal media supplemented with glucose, vitamins and amino acids with the exception of L-methionine. Contains base medium (500g), nutrient mix (120g), 250 X Selenomethionine (16mL @ 10mg/mL) and 250 X methionine. Enough to make 22L of media. 250 X Selenomethionine and 250 X methionine give you 4L. You have enough base medium and nutrient mix to make 22L of media. 250 X Selenomethionine and 250 X methionine give you 4L.
Catalog No. NC9695075