Learn More
Medchemexpress LLC Selenomethionine | 1464-42-2 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB100010MM/1ML
Encompass_Preferred
Selenomethionine is a naturally occurring amino acid containing selenium that has oral activity and is a common natural food source. It exhibits antitumor activity and has been studied for its potential to improve intervertebral disc degeneration.
  • Decreases hatchability and glutathione consumption
  • Inhibits growth of various cancer cells
  • Inhibits TBHP-induced oxidative stress
  • Prevents apoptosis of nucleus pulposus cells
  • Improves intervertebral disc degeneration in rat models
Catalog No. 50-004-07555