Learn More
Selleck Chemical LLC Seletalisib
SDP
Supplier:  Selleck Chemical LLC S85602MG
Encompass_Preferred
Seletalisib (UCB-5857) is a novel small-molecule inhibitor of PI3K with an IC50 value of 12 nM and shows significant selectivity to PI3K with respect to the other class I PI3K isoforms (between 24- and 303-fold)
Catalog No. 50-000-00698