Learn More
Medchemexpress LLC HY-14870 5mg , Selexipag CAS:475086-01-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY148705MG
Encompass_Preferred
HY-14870 5mg Selexipag CAS: 475086-01-2 Selexipag (NS-304) is an orally available and potent agonist for the Prostacyclin (PGI2) receptor (IP receptor). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0885