Learn More
GA International Self-Laminating Calibration Labels - "Calibration" 0.75" x 1.5" / 19.1mm x 38.1mm Rectangle Red text on White From -80°C to +93°C (-112°F to +199.4°F)
Supplier:  GA International CALA0011P
Encompass_Preferred
Self-laminating, writable, calibration labels designed for equipment that is calibrated or inspected regularly. The labels can be inscribed using permanent ink markers. The clear laminate is applied over the label protecting the written information. CalTAG™ labels can withstand a wide range of temperatures (-80°C to +93°C / -112°F to +200°F), abrasion and exposure to harsh chemicals, such as ethanol, acetone, isopropanol, thinners, detergents and surface sanitizing wipes.The self-laminating calibration labels are ideal for labeling regularly inspected medical equipment, equipment that has undergone preventative maintenance, that require calibration prior to use, or that are out of order and must not be used. Reagent calibration labels are also available for time-sensitive solutions prepared in the lab. Calibration labels customization is also available to include the company’s logos and information.
Catalog No. 50-005-71290