Learn More
Medchemexpress LLC HY-109137 100mg , Selgantolimod CAS:2004677-13-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY109137100MG
Encompass_Preferred
HY-109137 100mg Selgantolimod CAS: 2004677-13-6 Selgantolimod (GS-9688) is an orally active, potent and selective toll-like receptor 8 (TLR8) agonist for the treatment of hepatitis B virus (HBV) and human immunodeficiency virus (HIV) infection. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0509