Learn More
Selleck Chemical LLC Tirzepatide, 5 mg, synthetic dual agonist for GIP and GLP-1 receptors, powder

Supplier: Selleck Chemical LLC P12065MG
Tirzepatide (LY3298176) sodium is a high-purity, synthetic dual agonist for GIP and GLP-1 receptors, recognized for its effectiveness in promoting glucose-dependent insulin secretion and enhancing glucose tolerance. This compound is noted for its potential to aid in weight reduction and improving metabolic parameters.
- High purity: 99.96%
- 99% plasma protein binding to albumin
- Molecular weight: 4813 g/mol
- CAS number: 2023788-19-2
- Storage stability: 3 years at -20°C (powder), 1 year at -80°C (aliquoted stock solutions)
- Solubility in DMSO: 100 mg/mL (20.77 mM)
- Peptide length: 39 amino acids
- Chemical modification for improved stability and uptake
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.