Learn More
TARGETMOL CHEMICALS INC SELODENOSON 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T287485MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Selodenoson (RG-14202) is a selective adenosine A1 receptor agonist used to slow the heart rate in patients with atrial fibrillation and to treat arrhythmias. purity: 99%
Catalog No. 50-258-0497