Learn More
Medchemexpress LLC Selonsertib | 1448428-04-3 | 99.1% | 445.49 | 200 MG
SDP
Supplier:  Medchemexpress LLC HY18938200MG
Encompass_Preferred
Selonsertib (GS-4997) is an orally bioavailable, selective apoptosis signal-regulating kinase 1 (ASK1) inhibitor with a pIC50 of 8.3. It has been evaluated as an experimental treatment for diabetic nephropathy and kidney fibrosis. It is a highly selective and potent once-daily oral ASK1 inhibitor that competes with ATP in the ASK1 catalytic kinase domain.
  • Orally bioavailable.
  • Selective apoptosis signal-regulating kinase 1 (ASK1) inhibitor.
  • pIC50 of 8.3 for ASK1.
  • Competes with ATP in the ASK1 catalytic kinase domain.
  • Highly selective and potent once-daily oral ASK1 inhibitor.
  • Evaluated as an experimental treatment for diabetic nephropathy and kidney fibrosis.
Catalog No. 50-003-85184