Learn More
Selleck Chemical LLC Selonsertib
SDP
Supplier:  Selleck Chemical LLC S829225MG
Encompass_Preferred
Selonsertib (GS-4997) is a highly selective and potent once-daily oral ASK1 inhibitor with potential anti-inflammatory antineoplastic and anti-fibrotic activities
Catalog No. 50-000-00061