Learn More
Selleck Chemical LLC Selonsertib (GS-4997) 5mg 1448428-04-3
SDP
Supplier:  Selleck Chemical LLC S82925MG
Encompass_Preferred
Selonsertib (GS-4997) is a highly selective and potent once-daily oral ASK1 inhibitor with potential anti-inflammatory, antineoplastic and anti-fibrotic activities. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5910