Learn More
Medchemexpress LLC Seltorexant | 1293281-49-8 | 99.8% | 407.44 | 50 MG

Supplier: Medchemexpress LLC HY10901210MG
Seltorexant (JNJ-42847922) is an orally active, high-affinity, and selective orexin-2 receptor (OX2R) antagonist (pKi values of 8.0 and 8.1 for human and rat OX2R). It crosses the blood-brain barrier and quickly occupies OX2R binding sites in the rat brain.
- Target: Orexin-2 Receptor (OX2R)
- Activity: Orally active, high-affinity, and selective antagonist
- Brain permeability: Crosses the blood-brain barrier and rapidly occupies OX2R binding sites in the rat brain
- Purity: 99.83%
- Form: Solid
- Color: White to yellow
- Research use: Used for research purposes
- Sleep-promoting effects: Dose-dependently induces and prolongs sleep in male Sprague-Dawley rats, with effects maintained upon 7-day repeated dosing.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.