Learn More
Medchemexpress LLC Seltorexant (hydrochloride) | 1293284-49-7 | 99.9% | 443.91 | 5 MG
SDP
Supplier:  Medchemexpress LLC 5MGHY109012A
Encompass_Preferred
Seltorexant hydrochloride is an orally active, high-affinity, and selective OX2R antagonist. It has pKi values of 8.0 and 8.1 for human and rat OX2R, respectively. This compound crosses the blood-brain barrier and quickly occupies OX2R binding sites in the rat brain. It is intended for research use only.
  • Orally active, high-affinity, and selective OX2R antagonist
  • pKi values of 8.0 for human OX2R and 8.1 for rat OX2R
  • Crosses the blood-brain barrier
  • Quickly occupies OX2R binding sites in the rat brain
  • For research use only
Catalog No. 50-003-66005